Learn More
SSPE (20X), Thermo Scientific™
39.50 EUR - 104.00 EUR
Specifications
Concentration 20X
Physical Form Liquid
View More Specs
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15453829
View Documents
Thermo Scientific Alfa Aesar
J61214.K2
1 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15463829
View Documents
Thermo Scientific Alfa Aesar
J61214.K3
2 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15473829
View Documents
Thermo Scientific Alfa Aesar
J61214.K7
4 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
20X
Liquid
Videos
Safety and Handling
Safety and Handling
Recommended Storage : Ambient temperatures