Learn More
Square Unmounted Polarizing Filters
Brand:  N/A TYPE HN32
Additional Details : Weight : 0.00180kg
 View more versions of this product
Product Code. 12934717
  • POA / Pack of 2
Estimated Shipment: 01-08-2024
to see stock.
This item requires a price update prior to being ordered.
Request a Price for this item here.
Description
Description
Filters are supplied as a pair. For applications where highest optical quality is not essential. They are unmounted and may be cut with scissors.
Specifications
Specifications
Polarizing Filter
Lab Grade Plastic
For applications where highest optical quality is not essential.
50 x 50 mm