Learn More
Square Unmounted Polarizing Filters
Brand: N/A TYPE HN32
Additional Details : Weight : 0.00180kg
Description
Filters are supplied as a pair. For applications where highest optical quality is not essential. They are unmounted and may be cut with scissors.Specifications
Polarizing Filter | |
Lab Grade Plastic | |
For applications where highest optical quality is not essential. | |
50 x 50 mm |