missing translation for 'onlineSavingsMsg'
Learn More
Spermidine, free base, 99.5%, MP Biomedicals™ Product Code.: 11484371

Spermidine, free base, 99.5%, MP Biomedicals™

Product Code. 11484371 Shop All MP Biomedicals Products
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11484371

Brand: MP Biomedicals 0215206805

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 124-20-9
Molecular Formula C7H19N3
Molecular Weight (g/mol) 145.25
MDL Number MFCD00008229
InChI Key ATHGHQPFGPMSJY-UHFFFAOYSA-N
Synonym spermidine, 1,5,10-triazadecane, 4-azaoctamethylenediamine, spermidin, 4-azaoctane-1,8-diamine, n1-3-aminopropyl butane-1,4-diamine, 1,4-butanediamine, n-3-aminopropyl, n-3-aminopropyl butane-1,4-diamine, n-3-aminopropyl-1,4-butane-diamine, 1,4-diaminobutane, n-3-aminopropylShow MoreShow Less
PubChem CID 1102
ChEBI CHEBI:16610
IUPAC Name (4-aminobutyl)(3-aminopropyl)amine
SMILES NCCCCNCCCN

Specifications

Color Colorless
Density 0.925g/mL
Assay Percent Range ∽ 99%
Quantity 5 g
Formula Weight 145.25
Percent Purity 0.995
Physical Form Liquid
Chemical Name or Material Spermidine free base, Cell Biology and Analysis
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Spermidine, free base, 99.5%, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.