Learn More
asecos™ Sorbents Transport Trolley
Transport trolly for fast access in case of leakage.
Brand:  asecos™ 14413
 View more versions of this product
Product Code. 17827814
  • 681.00 EUR / Each
Estimated Shipment: 20-01-2025
Description
  • content: 6 cushions, 3 socks, 20 mats, 3 pair protective gloves, 5 plastic bags, 1 pair full vision safety glasses with elastic strap
  • 410 x 590 x 950 (W x D x H)
Specifications
Specifications
Grey
Spill Control Cart
16.14 in.
16.14 in.
37.40 in.
23.23 in.
77.16 lb.
Polyethylene
Special Sorbents
410.00 mm
410.00 mm
950.00 mm
590.00 mm
35 kg