missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium chloride, Free-flowing, Reagent Grade, anhydrous, ≥99%, Honeywell Fluka™
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
12 kg
25 kg
Packaging:
Fiberboard Box
Poly Bottle
Description
- ≥95% for general laboratory use.
Specifications
Specifications
Boiling Point | 1,465°C |
Melting Point | 801°C (literature) |
CAS Max % | ≤100.0000% |
Quantity | 2.5 kg |
Linear Formula | NaCl |
Beilstein | 3534976 |
IUPAC Name | sodium;chloride |
Formula Weight | 58.44 |
Percent Purity | ≥99% |
Grade | Reagent |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction