Learn More
Sodium chloride, Free-flowing, Reagent Grade, anhydrous, ≥99%, Honeywell Fluka™
Product Code. 15607120
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
12 kg
25 kg
Packaging:
Fiberboard Box
Poly Bottle
Free-flowing, Reagent Grade, anhydrous,≥99%
  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
SMILES [Na+].[Cl-]
Specifications
Boiling Point 1,465°C
Melting Point 801°C (literature)
CAS Max % ≤100.0000%
Quantity 1 kg
Linear Formula NaCl
Beilstein 3534976
IUPAC Name sodium chloride
Formula Weight 58.44
Percent Purity ≥99%
Grade Reagent
Packaging Poly Bottle
Density 2.160 g/cm3
Chemical Name or Material Sodium chloride, Free-flowing, Reagent Grade, anhydrous, ≥99%