Learn More
Sampling Systems™ Silicone Seals for Mini-Drums
These seals when used with toggle clamps will make mini-drum containers leak-proof. Choose the right sized seal for your mini-drum.
Brand:  Sampling Systems™ SIL-12-FDA
 View more versions of this product
Product Code. 15899721
  • 35.05 EUR / Each
Estimated Shipment: 25-02-2025
Description
  • O-ring design
  • Made from FDA-acceptable grade silicone
  • Suitable for use with both 304 and 316L stainless steel mini-drums
  • Wide range of sizes available
Specifications
Specifications
Leak-proof seals
12L mini-drum
Silicone
Seal, O-ring