Learn More
Cytiva Glass Plate
Plate made from glass.
Brand: Cytiva 80110629
Additional Details : Weight : 1.45000kg
Description
- For use with SDS and Native PAGE, IEF Casting Kit and NovaBlot kit
Specifications
SDS and Native PAGE, IEF Casting Kit and NovaBlot Kit | |
Glass Plate (125 x 260 x 1 mm) | |
Glass | |
15 |