Learn More
Epredia™ Shandon™ Tissue-Processing Capsule, Separators, S-shaped, Capsule
Partition up to four sections of the Epredia™ Shandon™ Tissue Processing Capsule, constructed of stainless steel.
Brand: Epredia™ 330
This item is not returnable.
View return policy
Description
- Five sturdy lugs grasp the base securely
- Partition separators sold separately
- Dimensions (O.D. x D): 1.46 x 0.24″ (37 x 6mm)