Learn More
3M™ SecureFit™ Anti-Scratch, Anti-Fog Safety Spectacles
Click to view available options
Lens Tint:
Amber
Blue Mirror
Clear
Gray
Indoor/Outdoor
Description
- Available in a range of lens tints
- Protection against UV radiation
- Naturally adjusts to individuals' head sizes
- Rimless lenses and fixed-length temple arm to enhance frame comfort
- Soft, adjustable nose pads for even greater personalization of fit
- Get added comfort over the ears with padded temple touchpoints
- Temple arms in a bold colour for an infusion of style
Specifications
Specifications
Lens Tint | Blue Mirror |
Product Type | Goggles |
No. per Pack | 1 Ea. |
Lens Material | Polycarbonate |
Promotions
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
Product Content Correction
Product Title
3M™ SecureFit™ Anti-Scratch, Anti-Fog Safety Spectacles >
Spot an opportunity for improvement?Share a Content Correction