Learn More
Masterflex™ One-Way Luer Check Valves
Connect these valves to bio-bags or sampling tubes. Cole-Parmer™ One-Way Luer Check Valves are molded onto the tubing for a seamless transition.
Brand: Masterflex™ 30505-92
Additional Details : Weight : 0.01080kg
Description
Valves feature low priming volume, ease of removing air bubbles during priming and operate in any spatial orientation. Manufactured DEHP-and latex-free silicone diaphragms. Materials meet USP CLass VI and ISO 10993 specifications. Each assembly comes packaged individually and can be sterilized.Specifications
One-Way Luer Fitting | |
SAN | |
10 Pack |