Learn More
Ruthenium, plasma standard solution, Specpure™ Ru 10,000μg/mL, Thermo Scientific Chemicals Product Code.: 11308677
Ruthenium, plasma standard solution, Specpure™ Ru 10,000μg/mL, Thermo Scientific Chemicals
Product Code. 11308677
100 mL
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Product Code. 11308677
Brand: Thermo Scientific Alfa Aesar 035773.AE
Request Bulk or Custom Format
Ruthenium, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7440-18-8
Molecular Formula Ru
Molecular Weight (g/mol) 101.07
MDL Number MFCD00011207 MFCD03458417
InChI Key KJTLSVCANCCWHF-UHFFFAOYSA-N
PubChem CID 23950
ChEBI CHEBI:30682
IUPAC Name ruthenium
SMILES [Ru]
Specifications
Quantity 100 mL
Physical Form Liquid
Solubility Miscible with water
Chemical Name or Material Ruthenium, Ru 10, 000μg/mL
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1526331 Certificate TypeCertificate of Analysis Date11/02/2025 Catalog Number035773.AD,035773.AE,035773.AP
Lot Number1508302 Certificate TypeCertificate of Analysis Date06/12/2024 Catalog Number035773.AD,035773.AE,035773.AP
Lot Number1508758 Certificate TypeCertificate of Analysis Date06/12/2024 Catalog Number035773.AD,035773.AE,035773.AP
Lot Number1507890 Certificate TypeCertificate of Analysis Date19/11/2024 Catalog Number035773.AD,035773.AE,035773.AP
Lot Number1490140 Certificate TypeCertificate of Analysis Date16/10/2024 Catalog Number035773.AD,035773.AE,035773.AP
Product Title
Ruthenium, plasma standard solution, Specpure™ Ru 10,000μg/mL, Thermo Scientific Chemicals >