Learn More
Thermo Scientific™ Replacement Seals for Thermo Scientific™ MicroClick and 8 x 8 PCR Strip Rotors
A range of options for secure closure on microcentrifuge rotors to help contain samples.
Brand:  Thermo Scientific™ 75005730
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15114933
  • 259.00 EUR / Each
Estimated Shipment: 21-06-2024
Description
A range of options for secure closure on microcentrifuge rotors to help contain samples.
  • O-rings for inner and outer seals
  • ClickSeal lids for optimal biocontainment
  • Available for a variety of sizes and models
Specifications
Specifications
Replacement ClickSeal Biocontainment Lid for MicroClick 30 x 2 and 8 x 8 PCR Strip Rotors