Learn More
Ohaus™ RAMP
Brand: Ohaus™ 22015466
675.04 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Order Info
LengthMetric
Specifications
Painted Steel | |
1250 mm | |
RAMP |
Product Content Correction
Product Title
Ohaus™ RAMP > Length: 1250mm
Spot an opportunity for improvement?Share a Content Correction