Learn More
Epredia™ RA Lamb Glass Coplin Jar, holds 5 slides (10 back to back)
Hold up to 10 slides back-to-back with Epredia™ RA Lamb molded glass coplin jars. Jars have a drop-on lid and integral large diameter base.
Brand: Epredia™ E94
Description
Features
- Heavy, clear glass construction with internal grooves
- 1.6 in. diameter circular tapered top
- Includes drop-on lid
Specifications
Yes | |
Round | |
Coplin Staining Jar | |
1.6 in. |
Drop-on lid | |
Glass | |
10 |