Learn More
Pyrex™ PTFE Liner for SciLabware 4526/45D Screw Cap
Brand: Pyrex 4546/45
This item is not returnable.
View return policy
Description
Specifications
45 mm | |
PTFE liner | |
10 Pack |
PTFE | |
PTFE |
Product Content Correction
Product Title
Pyrex™ PTFE Liner for SciLabware 4526/45D Screw Cap > Thread size: 45mm
Spot an opportunity for improvement?Share a Content Correction