Learn More
Cowie Technology™ PTFE Straight Through Stopcocks
Cowie Technology™ PTFE Straight Through Stopcocks feature a pure PTFE body and plug and combine all the desirable properties of the Ultimate Laboratory Stopcock.
14.20 EUR - 16.40 EUR
Specifications
Type | Straight Through |
---|---|
Description | Straight Through Stopcock |
Material | PTFE |
Autoclavable | Autoclavable |
No. per Pack | 1 Each |
Product Code | Brand | Plug Bore | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Plug Bore | Price | Quantity & Availability | |||||
10323552
|
Cowie Technology™
016.002 |
2 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10150192
|
Cowie Technology™
016.004 |
4 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10630613
|
Cowie Technology™
016.003 |
3 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Medium vacuum pressure performance
- Unbreakable
- Chemically inert
- Grease free
- Supplied with borosilicate glass sidearms
Specifications
Straight Through | |
PTFE | |
1 Each | |
PTFE |
Straight Through Stopcock | |
Autoclavable | |
Borosilicate glass sidearms |