Learn More
Cole-Parmer™ PTFE Sterile Syringe Filters
Unaffected by most organic solvents, concentrated acids and bases, propellants and cryogenic fluids. Polytetrafluoroethylene (PTFE) membranes are ideal for sterilization of gas and alcohols; cleaning acids, alkalies, and organic solvents; venting purposes.
Brand: Cole-Parmer™ 02915-08
Additional Details : Weight : 0.29000kg
Description
- Filters are Triton-free and have a bidirectional membrane support and luer lock (15/25 mm dia.) or stepped barbs (50mm dia.) inlet/outlet for a secure fit to the syringe
- Membrane type and pore size are printed on each filter for easy identification
- All Cole-Parmer syringe filters are 100% integrity tested and manufactured in accordance with ISO 9002 standards
- Sterile filters are sterilized by gamma radiation
- Certified nonpyrogenic and noncytotoxic
- Blister packed
Specifications
Natural | |
PTFE | |
Sterilization of gas and alcohols; cleaning acids, alkalies and organic solvents | |
50 Filters | |
Sterile Syringe Filters | |
25 mm |
PTFE/Polypropylene | |
0.2 μm | |
Polypropylene | |
Retention: ≥0.35mL | |
100 mL |