Learn More
Potassium Sulfate, ≥99.0%, ACS Reagent, Honeywell Fluka™
Product Code. 15612220
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
ACS Reagent,≥99.0%, powder
  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7778-80-5
Molecular Formula K2O4S
Molecular Weight (g/mol) 174.25
MDL Number MFCD00011388
InChI Key OTYBMLCTZGSZBG-UHFFFAOYSA-L
Synonym potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
PubChem CID 24507
ChEBI CHEBI:32036
IUPAC Name dipotassium sulfate
SMILES [K+].[K+].[O-]S([O-])(=O)=O
Specifications
Boiling Point 1698°C
Melting Point 1067°C
pH 6.0 to 7.5 at 20°C
Physical Form Powder
Quantity 2.5 kg
Linear Formula K2SO4
UN Number NONH for all modes of transport
Formula Weight 174.26g/mol
Percent Purity ≥99.0%
Grade ACS Reagent
Packaging Poly Bottle
Appearance Powder
Chemical Name or Material Potassium Sulfate