Learn More
Potassium Phosphate Monobasic Anhydrous, MP Biomedicals
35.55 EUR - 550.00 EUR
Chemical Identifiers
CAS | 7778-77-0 |
---|---|
Molecular Formula | H2KO4P |
Molecular Weight (g/mol) | 136.08 |
MDL Number | MFCD00011401 MFCD00147253 |
InChI Key | GNSKLFRGEWLPPA-UHFFFAOYSA-M |
Synonym | potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphateShow More |
PubChem CID | 516951 |
ChEBI | CHEBI:63036 |
IUPAC Name | potassium dihydrogen phosphate |
SMILES | [K+].OP(O)([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11430562
![]() |
MP Biomedicals
02191430.1 |
100 g |
35.55 EUR
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11450562
![]() |
MP Biomedicals
0219143001 |
1 kg |
123.00 EUR
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11460562
![]() |
MP Biomedicals
0219143005 |
5 kg |
550.00 EUR
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
7778-77-0 | |
136.08 | |
GNSKLFRGEWLPPA-UHFFFAOYSA-M | |
516951 | |
potassium dihydrogen phosphate |
H2KO4P | |
MFCD00011401 MFCD00147253 | |
potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphateShow More | |
CHEBI:63036 | |
[K+].OP(O)([O-])=O |
Specifications
7778-77-0 | |
MFCD00011401 MFCD00147253 | |
GNSKLFRGEWLPPA-UHFFFAOYSA-M | |
potassium dihydrogen phosphate | |
516951 |
H2KO4P | |
potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate | |
[K+].OP(O)([O-])=O | |
136.08 | |
CHEBI:63036 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Potassium Phosphate Monobasic Anhydrous, MP Biomedicals