missing translation for 'onlineSavingsMsg'
Learn More

Potassium phosphate, 0.2M buffer soln., pH 7.6, Thermo Scientific™

50.80 EUR - 67.90 EUR

Specifications

CAS 7778-77-0
Concentration 0.2 M
pH 7.6
Physical Form Liquid
Molecular Formula H2KO4P
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15415399
View Documents
Thermo Scientific Alfa Aesar
J63128.AP
500 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15425399
View Documents
Thermo Scientific Alfa Aesar
J63128.K2
1 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Potassium phosphate buffer is typically used as a component for a wide variety of media used in the culture of microorganisms, as a component in phosphate buffered saline (PBS). In addition to helping maintain pH, it supplies essential phosphate.

Solubility
Soluble in water, Insoluble to slightly soluble in ethanol, insoluble in alcohol.

Notes
Store sealed bottle containing the dehydrated medium at 2 - 30C. Once opened and recapped, place container in a low humidity environment at the same storage temperature. Protect from moisture and light by keeping container tightly closed.
Specifications

Specifications

7778-77-0
7.6
H2KO4P
GNSKLFRGEWLPPA-UHFFFAOYSA-M
potassium dihydrogen phosphate
0.2 M
Liquid
MFCD00011401 MFCD00147253
[K+].OP(O)([O-])=O
136.08
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Ambient Temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium phosphate, 0.2M buffer soln., pH 7.6, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.