Learn More
Potassium Permanganate, Ph. Eur. Grade, 0.02M, Reagecon™
BIOPHARMA
Product Code. 11814403
Click to view available options
Quantity:
1 L
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
Specifications
Quantity 1 L
compliance-icons
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
  • P391-Collect spillage.
Supplemental information
  • MIXTURE LIST-Contains : Potassium Permanganate 99%