Learn More
Potassium Bromate, Extra Pure, SLR, Fisher Chemical™
Product Code. 10715151
Click to view available options
Quantity:
250 g
500 g
Chemical Identifiers
CAS 7758-01-2
Molecular Formula BrKO3
Molecular Weight (g/mol) 167.00
MDL Number 11359
InChI Key OCATYIAKPYKMPG-UHFFFAOYSA-M
Synonym potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat
PubChem CID 23673461
ChEBI CHEBI:38211
IUPAC Name potassium;bromate
SMILES [O-]Br(=O)=O.[K+]
Specifications
Melting Point 350°C
Color White
pH 5 to 9
Physical Form Powder/Solid
Quantity 500 g
Formula Weight 167.01g/mol
Packaging Plastic powder jar