Learn More
Cole-Parmer™ Polystyrene Storage Container
Brand:  Cole-Parmer™ 06816-41
Product Code. 15248898
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Specifications
Polystyrene
Storage Container
1.6 mm
Rectangular
24 Pack
1.9 mm
Documents
One moment while we fetch your results.
Product Title
Cole-Parmer™ Polystyrene Storage Container > Height: 1.9mm