Learn More
Kartell™ Ranvier Type Dropping Bottle
Click to view available options
Capacity (Metric):
50 mL
100 mL
Description
- Comes with PE tube and rubber teat
- Suitable for foodstuff
Specifications
Specifications
Capacity (Metric) | 50 mL |
Color | Neutral |
For Use With (Application) | Foodstuff |
Includes | Polyethylene tube, Rubber teat |
Material | Polyethylene |
Quantity | 10Pack |
Product Line | Bottle |
Height (Metric) | 96 mm |
Product Type | Dropping Bottle |
Diameter (Metric) | 35 mm |
Product Content Correction
Product Title
Kartell™ Ranvier Type Dropping Bottle >
Spot an opportunity for improvement?Share a Content Correction