Learn More
Thermo Scientific™ Point of Dispense Filters for Barnstead™ Point of Use Water Purification Systems
Sterile filter for use with Barnstead™ water purification systems; can be autoclaved up to 5 times.
221.00 EUR - 270.00 EUR
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15950206
|
Thermo Scientific™
50157375 |
Sterile Filter, 0.1μm |
270.00 EUR
1 unit |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
11367111
|
Thermo Scientific™
09.1003 |
Sterile Filter, 0.2μm |
221.00 EUR
Each |
In Stock
Log in to see stock. |
|||||
Description
Reduce bacteria using the Thermo Scientific™ Barnstead™ Point of Use Water Purification Systems.
Replacement consumable for the following Barnstead water purification systems:
- GenPure™
- GenPure™ Pro
- GenPure™ xCAD
- LabTower™ EDI
- LabTower™ TII
- MicroPure™
- Smart2Pure™
- Smart2Pure™ Pro