Learn More
Memmertā„¢ Plug-In Tube Extension for UF160, Straight
Plug-in tube extension designed for exhaust air ducting and connection by hose if necessary.
Brand:  Memmertā„¢ UF160B29718
Product Code. 17559005
  • 51.25 EUR / Each
Only null left
Estimated Shipment: 21-07-2025
Description
  • Accessory
  • Plug-in tube extension (outer diameter 60.3 mm, inner 57 mm)
  • Straight
Specifications
Specifications
UF160 Oven
Plug-in Tube Extension
O.D. 60.3 mm, I.D. 57 mm
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Memmertā„¢ Plug-In Tube Extension for UF160, Straight > For Use With: UF160