Learn More
Memmert™ Plug-In Tube Extension (Od/Id 60.3/57 mm), Angled, For Exhaust Air Ducting For S260
Use these tube extensions with Memmert ovens. Memmert™ Plug-in Tube Extensions for Ovens are designed for exhaust air ducting and connection by hose if necessary.
Brand: Memmert™ S260B29719
This item is not returnable.
View return policy
Description
- Plug-in tube extension (outer diam. 60,3 mm, inner 57 mm)
- Angled
- Only models U, I, S
Specifications
Exhaust Air Ducting | |
Plug-In Tube Extension | |
57 x 60.3 mm (I.D. x O.D.) |