Learn More
Phosphotungstic acid hydrate, 83.51%, MP Biomedicals™
141.00 EUR - 321.00 EUR
Chemical Identifiers
CAS | 12501-23-4 |
---|---|
Molecular Formula | H5O41PW12 |
Molecular Weight (g/mol) | 2880.20 |
MDL Number | MFCD00011342 |
InChI Key | AVFBYUADVDVJQL-UHFFFAOYSA-N |
Synonym | phosphotungstic acid hydrate, tungstophosphoric acid, 12-tungstophosphoric acid hydrate, phosphoric acid dodecakis tungsten trioxide hydrate, iuaxea, aronis24073, phosphotungstic acid hydrate, reagent grade, phosphotungstic acid hydrate, for microscopy, phosphotungstic acid hydrate, p.a, phosphotungstic acid hydrate, saj special grade |
PubChem CID | 16212977 |
IUPAC Name | 12-Tungstophosphate hydrate |
SMILES | O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.OP(O)(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11463612
|
MP Biomedicals
0215624380 |
100 g |
141.00 EUR
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11473612
|
MP Biomedicals
0215624383 |
250 g |
321.00 EUR
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
12501-23-4 | |
2880.20 | |
AVFBYUADVDVJQL-UHFFFAOYSA-N | |
16212977 | |
O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.OP(O)(O)=O |
H5O41PW12 | |
MFCD00011342 | |
phosphotungstic acid hydrate, tungstophosphoric acid, 12-tungstophosphoric acid hydrate, phosphoric acid dodecakis tungsten trioxide hydrate, iuaxea, aronis24073, phosphotungstic acid hydrate, reagent grade, phosphotungstic acid hydrate, for microscopy, phosphotungstic acid hydrate, p.a, phosphotungstic acid hydrate, saj special grade | |
12-Tungstophosphate hydrate |
Specifications
12501-23-4 | |
Crystalline Powder | |
phosphotungstic acid hydrate, tungstophosphoric acid, 12-tungstophosphoric acid hydrate, phosphoric acid dodecakis tungsten trioxide hydrate, iuaxea, aronis24073, phosphotungstic acid hydrate, reagent grade, phosphotungstic acid hydrate, for microscopy, phosphotungstic acid hydrate, p.a, phosphotungstic acid hydrate, saj special grade | |
O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.OP(O)(O)=O | |
2880.20 | |
2880.2 | |
Phosphotungstic acid hydrate |
H5O41PW12 | |
MFCD00011342 | |
AVFBYUADVDVJQL-UHFFFAOYSA-N | |
12-Tungstophosphate hydrate | |
16212977 | |
83.51% |
Safety and Handling
Recommended Storage : Store at Room Temperature.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Phosphotungstic acid hydrate, 83.51%, MP Biomedicals™