Learn More
Fisherbrand™ Polystyrene Petri Dishes, Sterile
A range of high quality aseptic or sterile Petri dishes perfect for microbial or cell culture, or for sample collection, examination and storage.
Brand: Fisherbrand™ FB51510
This item is not returnable.
View return policy
Description
Specifications
Polystyrene | |
Petri Dish | |
3 | |
90 mm |
Sterile | |
600 Case | |
16.2 mm |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
M552140 | Certificate of Compliance | 17/12/2024 | FB51510 |
Product Content Correction
Product Title
Fisherbrand™ Polystyrene Petri Dishes, Sterile > 3 Vents; Height: 16.2mm
Spot an opportunity for improvement?Share a Content Correction