Learn More
Palladium Silver foil, 0.025mm (0.001in) thick Product Code.: 15473915
Palladium Silver foil, 0.025mm (0.001in) thick
Product Code. 15473915
25 x 25 mm, Each
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
Unit Size:
Each
Product Code. 15473915
Brand: Thermo Scientific Alfa Aesar 045003.FF
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Palladium Silver foil
Name Note 0.025mm (0.001 in.) thick
Avg. Mol. Wt. or Mol. Wt. Range Pd:Ag; 77:23 wt%
Assay Percent Notes (metals basis excluding Pt)
MDL Number MFCD02091738
Quantity 25 x 25 mm
Percent Purity 99.9%
Odor Odorless
Weight ≈0.18g/25x25mm
Form Foil
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberR29L016 Certificate TypeCertificate of Analysis Date12/06/2025 Catalog Number045003.GH,045003.FF,045003.FI
Lot NumberH07T026 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog Number045003.FF,045003.FI
Product Title
Palladium Silver foil, 0.025mm (0.001in) thick >