Learn More
p-Tolylmagnesium bromide, approx. 0.5M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals
p-Tolylmagnesium bromide, approx. 0.5M solution in diethyl ether, AcroSeal, Acros Organics, Quantity: 800mL, Packaging: AcroSeal Glass bottle, Molecular Weight: 195.34, CAS: 4294-57-9, 60-29-7, Density: 0.8660g/mL, Flash Point: -40 deg.C, Formula Weight: 195.34
61.81 EUR - 329.00 EUR
Chemical Identifiers
CAS | 4294-57-9, 60-29-7 |
---|---|
Molecular Formula | C7H7BrMg |
Molecular Weight (g/mol) | 195.34 |
MDL Number | MFCD00000040 |
InChI Key | ZRJNGFJIBZKXTP-UHFFFAOYSA-M |
Synonym | p-tolylmagnesium bromide, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide, grignard reagent |
PubChem CID | 3644322 |
IUPAC Name | magnesium;methylbenzene;bromide |
SMILES | CC1=CC=C([Mg]Br)C=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10433464
|
Thermo Scientific Acros
426761000 |
100 mL | AcroSeal™ Glass bottle |
61.81 EUR
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10683395
|
Thermo Scientific Acros
426768000 |
800 mL | AcroSeal™ Glass bottle |
329.00 EUR
800mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4294-57-9, 60-29-7 | |
195.34 | |
ZRJNGFJIBZKXTP-UHFFFAOYSA-M | |
3644322 | |
CC1=CC=C([Mg]Br)C=C1 |
C7H7BrMg | |
MFCD00000040 | |
p-tolylmagnesium bromide, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide, grignard reagent | |
magnesium;methylbenzene;bromide |
Specifications
4294-57-9, 60-29-7 | |
-40°C | |
CH3C6H4MgBr | |
0.866 | |
ZRJNGFJIBZKXTP-UHFFFAOYSA-M | |
magnesium;methylbenzene;bromide | |
3644322 | |
p-Tolylmagnesium bromide |
0.8660g/mL | |
C7H7BrMg | |
MFCD00000040 | |
p-tolylmagnesium bromide, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide, grignard reagent | |
CC1=CC=C([Mg]Br)C=C1 | |
195.34 | |
195.34 |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Harmful if swallowed.
Extremely flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
- p-Tolylmagnesium bromide, ca. 0.4M(10%) solution in diethyl ether
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
- H224-Extremely flammable liquid and vapour.
- EUH014-Reacts violently with water.
- EUH019-May form explosive peroxides.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P243-Take precautionary measures against static discharge.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
- P403+P235-Store in a well-ventilated place. Keep cool.