Learn More
Thermo Scientific™ Oxoid™ Antimicrobial Susceptibility disc Dispenser
Accurately place antimicrobial susceptibility discs onto your media of choice when carrying out manual Antimicrobial Susceptibility Testing methods.
Brand: Thermo Scientific™ ST8100
527.25 EUR valid until 2024-05-31
Use promo code "21654" to get your promotional price.
Additional Details : Weight : 1.00000kg
Description
Accurately place antimicrobial susceptibility discs onto your media of choice when carrying out manual Antimicrobial Susceptibility Testing methods using a self-tamping Thermo Scientific™ Oxoid™ Antimicrobial Susceptibility disc Dispenser.
One of the simplest and most widely used methods for susceptibility testing, disc diffusion is extremely flexible, offering a wide variety of antibiotics and concentrations which can easily be changed as required.disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results but also provides a visual indication of the following:
- Inoculum level
- Presence of contamination
- Resistant mutants
- Beta-lactamase activity
- Antagonism and synergy between antibiotics
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
For use only with Oxoid Antimicrobial Susceptibility Discs | |
Antimicrobial Susceptibility Disc Dispenser | |
8 Oxoid Disc Cartridges |
Oxoid Antimicrobial Susceptibility Disc Dispenser | |
100mm diameter plates | |
1 Dispenser/Pk. |
For use only with Oxoid™ Antimicrobial Susceptibility discs.