Learn More
Thermo Scientific™ A2 and A5 Large Gel System Replacement End Gates
Use these gasketed UVT gel trays, replacement gaskets and end gates with Thermo Scientific™ A2 and A5 Large Gel Systems.
66.10 EUR
Specifications
For Use With (Equipment) Owl A2 and A5 Large Gel Electrophoresis System
View More Specs
Products 1
Product Code Brand Type Price Quantity & Availability  
Product Code Brand Type Price Quantity & Availability  
12463657
View Documents
Thermo Scientific™
A2-EG
End Gates
66.10 EUR
Each
Only null left
Estimated Shipment: 21-08-2025
Add to basket
Description
Description
Use Thermo Scientific™ A2 and A5 Large Gel System Replacement Parts with Owl A2 and A5 Large Gel Systems.
  • UVT gel tray incorporates gasketed end gates which provide a leakproof seal without tape
  • Comb slots are positioned to allow 2, 3 or 4 sample sets to run equal lengths on the same gel
Specifications
Specifications
Owl A2 and A5 Large Gel Electrophoresis System
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ A2 and A5 Large Gel System Replacement End Gates