Learn More
Thermo Scientific™ Orion™ Electrode Adapter Cables
Convert an electrode with one connector to the required connector of another meter by using the Thermo Scientific™ Orion™ Electrode Adapter Cables.
Brand: Thermo Scientific™ 090032
Additional Details : Weight : 1.36080kg
Description
Convert an electrode with one connector to the required connector of another meter by using the Thermo Scientific™ Orion™ Electrode Adapter Cables.
- For use with meters that have BNC, US standard, certain LEMO, British, E DIN, Radiometer, 2mm reference pins or just a stripped lead inputs
- Electrodes must have a US standard, screw cap, Karl Fisher, or BNC connectors
Specifications
BNC | |
Meters That Have BNC | |
U.S. Standard |
U.S. Standard; BNC; BNC to U.S. Standard Adapter | |
Electrode Cable |