Learn More
Cytiva Omega Membrane Discs
Omega Membrane Discs with high performance and stability for users of conventional, non-disposable stirred cells.
Brand:  Cytiva OM100043
 View more versions of this product
Product Code. 17114821
  • 376.00 EUR / Pack of 12
Only null left
Estimated Shipment: 09-10-2025
Description
  • Polyethersulfone membrane provides higher flow rates and lower protein binding than competitive regenerated cellulose
  • Lower processing times and the highest possible recoveries
  • Discs can be washed and reused
  • Replacement membranes for other stirred cell systems concentration, fractionation, purification, buffer exchange, or desalting of protein, cell broths, and other biomolecules
  • For research use only
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PES
Non-sterile
43 mm
100 kDa
12/Pk.
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Cytiva Omega Membrane Discs > 100 kDa; 43 mm