Learn More
Thermo Scientific Alfa Aesar NICKEL PLATING SOLUTION ELECTROLESS 1L
Brand:  Thermo Scientific Alfa Aesar 044215.K2
NICKEL PLATING SOLUTION ELECTROLESS 1L
Product Code. 11349728
  • 95.10 EUR / 1 L
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific Alfa Aesar NICKEL PLATING SOLUTION ELECTROLESS 1L >