Learn More
Thermo Scientific™ Nalgene™ Torque Wrench Fittings for ETFE Closures
Assure accurate torque readings when applying closures onto bottles with Nalgene™ Torque Wrench Fittings for ETFE closures.
129.00 EUR - 285.00 EUR
Specifications
Product Type | Fittings |
---|---|
Drive Size | 1/4 in. |
For Use With (Equipment) | Nalgene Torque Wrenches (Cat. No. 2195-6169) |
Material | Epoxy |
No. per Pack | 1 Ea. |
Product Code | Brand | Closure Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Size | Price | Quantity & Availability | |||||
11994932
|
Thermo Scientific™
2195-2020 |
20 to 415 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11904942
|
Thermo Scientific™
2195-2024 |
24 to 415 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11914942
|
Thermo Scientific™
2195-2028 |
28 to 415 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11924942
|
Thermo Scientific™
2195-2033 |
33 to 415 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11934942
|
Thermo Scientific™
2195-2038 |
38 to 415 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11944942
|
Thermo Scientific™
2195-2043 |
43 to 415 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11954942
|
Thermo Scientific™
2195-2048 |
48 to 415 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11964942
|
Thermo Scientific™
2195-2053 |
53 to 415 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11974942
|
Thermo Scientific™
2195-2438 |
38 to 430 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Assure accurate torque readings when applying closures onto bottles with Thermo Scientific™ Nalgene™ Torque Wrench Fittings for ETFE Closures. Proper application torque is essential for leakproof† performance.
- Durable epoxy resin construction
- Specifically designed for Nalgene ETFE closures
Compatible with:
Compatible with Thermo Scientific Nalgene torque wrench 2195-6169 † The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Warranty: 90 days
Specifications
Fittings | |
Nalgene Torque Wrenches (Cat. No. 2195-6169) | |
1 Ea. |
1/4 in. | |
Epoxy |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.