missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(acryloyl)cystamine, 98%, Thermo Scientific Chemicals

Catalog Number p-4408317
Click to view available options
Quantity:
1 g
5 g
25 g
60984-57-8
C10H16N2O2S2
260.37
MFCD00036225
DJVKJGIZQFBFGS-UHFFFAOYSA-N
n,n'-bis acryloyl cystamine, n,n'-bisacrylylcystamine, 2-propenamide, n,n'-dithiodi-2,1-ethanediyl bis, 2,2'-bisacrylamino diethyldisulfide, bis-acrylylcystamine: bac, n,n'-dithiodi-2,1-ethanediyl bis acrylamide, n-2-2-prop-2-enamido ethyl disulfanyl ethyl prop-2-enamide, n-2-2-prop-2-enoylamino ethyldisulfanyl ethyl prop-2-enamide, acmc-20ak9p, n,n-bis-acrylylcystamine
100602
N-[2-[2-(prop-2-enoylamino)ethyldisulfanyl]ethyl]prop-2-enamide
C=CC(=O)NCCSSCCNC(=O)C=C
60984-57-8
C10H16N2O2S2
260.37
MFCD00036225
DJVKJGIZQFBFGS-UHFFFAOYSA-N
n,n'-bis acryloyl cystamine, n,n'-bisacrylylcystamine, 2-propenamide, n,n'-dithiodi-2,1-ethanediyl bis, 2,2'-bisacrylamino diethyldisulfide, bis-acrylylcystamine: bac, n,n'-dithiodi-2,1-ethanediyl bis acrylamide, n-2-2-prop-2-enamido ethyl disulfanyl ethyl prop-2-enamide, n-2-2-prop-2-enoylamino ethyldisulfanyl ethyl prop-2-enamide, acmc-20ak9p, n,n-bis-acrylylcystamine
100602
N-[2-[2-(prop-2-enoylamino)ethyldisulfanyl]ethyl]prop-2-enamide
C=CC(=O)NCCSSCCNC(=O)C=C

Used as a reversible cross-linker for polyacrylamide gels

N,N'-Bis(acryloyl)cystamine is used as a reversible cross-linker for polyacrylamide gels. It is also used for DNA purification. Further, it finds application in Suzuki reaction and fabrication of hydrogel modified plasmonic crystal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 123°C to 124°C
Assay Percent Range 98%
Quantity 1 g
Solubility Information Soluble in acetic acid. Slightly soluble in water.
Formula Weight 260.37
Physical Form Powder
Chemical Name or Material N, N′-Bis(acryloyl)cystamine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'