missing translation for 'onlineSavingsMsg'
Learn More
Muscone, ≥95%, MP Biomedicals™ Product Code.: 11481391

Muscone, ≥95%, MP Biomedicals™

Product Code. 11481391 Shop All MP Biomedicals Products
500 mg
Click to view available options
Quantity:
250 mg
500 mg
This item is not returnable. View return policy

Product Code. 11481391

Brand: MP Biomedicals 0215918690

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Originally collected from male musk deer ventral glands, is a potent antiinflammatory agent that has been reported to reduce proinflammatory cytokines in vitro

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 541-91-3
Molecular Formula C16H30O
Molecular Weight (g/mol) 238.42
MDL Number MFCD00211114
InChI Key ALHUZKCOMYUFRB-UHFFFAOYNA-N
Synonym muscone, 3-methylcyclopentadecanone, muskone, methylexaltone, moschus ketone, cyclopentadecanone, 3-methyl, 3-methyl-1-cyclopentadecanone, 5-methyl-1-cyclopentadecanone, fema no. 3434, 3-methyl-cyclopentadecanoneShow MoreShow Less
PubChem CID 10947
IUPAC Name 3-methylcyclopentadecan-1-one
SMILES CC1CCCCCCCCCCCCC(=O)C1

Specifications

Color Colorless
Density 0.84g/mL
Assay Percent Range ≥95%
Quantity 500 mg
Formula Weight 238.43
Physical Form Liquid
Chemical Name or Material Muscone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Muscone, ≥95%, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.