Learn More
Fisherbrand™ Plan Achromatic Microscope Objective PH+
Correct for color while using an inverted microscope.
Brand: Fisherbrand™ 3623.0259
This item is not returnable.
View return policy
Description
- For use with inverted microscopes
- Corrects for color
-
Specifications
Plan Achromatic Objective | |
25x/0.40/160/1.2 LWD PH+ | |
Microscope model: FB69196 | |
25X |