Learn More
Micronova™ CGT Conductive Grid Tape
This conductive, anti-static polypropylene tape is specially designed with conductive grids embedded between two anti-static layers, for use on sensitive devices in ESD sensitive environments.
Brand:  Micronova™ CGT-1
 View more versions of this product
Product Code. 12837023
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
The Micronova CGT Conductive Grid Tape is a heavy duty, anti-static copolymer tape with acrylic adhesive. The conductive grids are embedded between two anti-static layers that provide an effective static shield, preventing static build-up, as well as preventing any shedding, flaking, cracking or chipping of the grids.
  • Heavy duty, anti-static copolymer tape with acrylic adhesive
  • Manufactured for use in ESD sensitive environments
  • Features a conductive grid embedded between two anti-static layers
  • Protects sensitive devices that would be otherwise damaged by normal insulating tape
  • Provides an effective static shield and prevents static buildup
  • Prevents shedding, flaking, cracking or chipping of the grids
  • 36 yards wound on a 3 in. plastic core
Order Info
Roll length, 36 yd. (33m)
Specifications
Specifications
Not Autoclavable
Clear
Conductive Grid Cleanroom Tape
Sealing in ESD Sensitive Environment-Conducts Charge Away From Substrate
16°C to 27°C
36 yd.
1 in.
Acrylic
Polypropylene
Non-sterile
60°F to 80°F
36 yd. x 1 in.
33 m
25.4 mm
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number047779-000 Certificate TypeCertificate of Compliance Date31/01/2025 Catalog NumberCGT-1
Lot Number043875-000 Certificate TypeCertificate of Compliance Date03/06/2024 Catalog NumberCGT-1
Lot Number043848-000 Certificate TypeCertificate of Compliance Date20/05/2024 Catalog NumberCGT-1
Lot Number040179-000 Certificate TypeCertificate of Compliance Date21/09/2023 Catalog NumberCGT-1
Lot Number036715-000 Certificate TypeCertificate of Compliance Date09/05/2023 Catalog NumberCGT-1
Promotions
Promotions
Product Content Correction
Product Title
Micronova™ CGT Conductive Grid Tape > CGT Conductive Grid Tape, anti-static co-polymer, 3 in. plastic core, 1 in. wide