Learn More
Metro™ MetroMax™ iQ Polymer Shelving Accessory, Color Shelf Marker
Use to assign color coding to individual shelf levels, shelving units, carts, or strorage areas
Brand:  Metro™ CSM6-RX
Additional Details : Weight : 0.02400kg
 View more versions of this product
Product Code. 15926901
  • 7.40 EUR / Each
Estimated Shipment: 03-10-2024
Description
  • Attaches easity to shelf for content identification
  • 6 in. length
Specifications
Specifications
6 in.
Metro™ MetroMax i™ and Q™ shelving
15.2 cm