missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Methyl Green, zinc chloride Product Code.: 15404109

Thermo Scientific Chemicals Methyl Green, zinc chloride

Product Code. 15404109
100 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 15404109

Brand: Thermo Scientific Chemicals J61509.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7114-03-6 | C27H35Cl4N3Zn | 608.78 g/mol

Methyl Green is a DNA stain used as a tracking dye in acid buffers for electrophoresis. It helps differentiate bacteria and is also a tissue stain for nucleoli and mitochondria in tissues. It is used with Bismarck Brown frequently.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl Green is a DNA stain used as a tracking dye in acid buffers for electrophoresis. It helps differentiate bacteria and is also a tissue stain for nucleoli and mitochondria in tissues. It is used with Bismarck Brown frequently.

Solubility
Soluble in water, ethanol or methanol

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7114-03-6
Molecular Formula C27H35Cl4N3Zn
Molecular Weight (g/mol) 608.78
MDL Number MFCD00151094
InChI Key ZKOGXCSOLMZRAZ-UHFFFAOYSA-J
Synonym C.I. 42590
PubChem CID 131675247
IUPAC Name zinc;[4-[[4-(dimethylamino)phenyl]-(4-dimethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]phenyl]-ethyl-dimethylazanium;bromide;trichloride
SMILES CC[N+](C)(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-].[Cl-].[Cl-].[Zn+2].[Br-]

Specifications

Melting Point >300°C
Beilstein 3914106
Solubility Information Soluble in water,ethanol or methanol
Formula Weight 653.24
Color Green
Quantity 100 g
Physical Form Powder
Chemical Name or Material Methyl Green, zinc chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Methyl Green, zinc chloride >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.