Learn More
Cole-Parmer™ Mesh Shelf
Static stainless steel Mesh Shelf for use in Techne hybridizers, able to support up to 4 kg in weight.
195.00 EUR - 202.00 EUR
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
12823386
|
Cole-Parmer™
39455-26 |
HB-1D Hybridizer |
202.00 EUR
Each |
Estimated Shipment: 27-03-2025
Log in to see stock. |
|||||
11563849
|
Cole-Parmer™
39455-22 |
HB-3D Hybrigene |
195.00 EUR
Each |
Estimated Shipment: 27-03-2025
Log in to see stock. |
|||||
Description
- Can be placed into the tube locators.
- Allows the unit to be used as a simple non-motion incubator.
- For use with either HB-1D or HB-3D hybridizers.
Specifications
Up to 4 kg | |
Stainless Steel | |
Shelf |