missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melanin, MP Biomedicals™
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
Specifications
Specifications
Color | Brown to Black |
Quantity | 250 mg |
Formula Weight | 1780 |
Physical Form | Granular Powder |
Chemical Name or Material | Melanin |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction