Learn More
Motic™ MB Filter Cube, BA310E
For detection of methylene blue. Motic™ MB Filter Cube, BA310E is suitable for BA310E Microscopes.
Brand:  Motic™ 1101001902925
Additional Details : Weight : 0.01000kg
Product Code. 15405252
  • 2000.00 EUR / Each
Estimated Shipment: 01-08-2024
Description
  • Epi-LED fluorescence equipment for methylene blue.
  • Cube houses an excitation filter, an emission filter, and a dichroic beam splitter.
  • Suitable for BA-310E Microscopes.
Specifications
Specifications
Filter Cube, MB
BA-310E Microscopes
Excitation Filter, Emission Filter, Dichroic Beam splitter