Learn More
OHAUS Mat, Sticky, 22 x 18 cm
Endeavor 5000 Light Duty Shaker Accessory
Brand:  OHAUS 30770939
71.20 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 17941282
  • 76.90 EUR / Each
Only null left
Estimated Shipment: 07-07-2025
Description
  • 22 x 18 cm
shaker, orbital, digital, endeavor, mat, sticky
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mat
Microbiology, Molecular Biology, ELISA Assays, Cell Cultures, Immunoassays, Protein Studies, Bacterial and Yeast Cultures, Staining/Destaining, Blotting Techniques, DNA Studies, Solubility Studies, Environmental and Food Sciences, Chemistry, General Mixing
Certificates
Promotions
Promotions
Product Content Correction
Product Title
OHAUS Mat, Sticky, 22 x 18 cm > Product Type: Mat