Learn More
BRAND™ Soda Lime Glass Culture Tubes with Screw Cap
Manufactured from soda lime glass. BRAND™ Soda Lime Glass Culture Tubes with Screw Cap include a polypropylene screw cap with white elastomer seal and GL thread. Autoclavable at 121°C.
Brand: BRAND™ 113941
89.59 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Description
- Manufactured from soda lime glass.
- Includes polypropylene screw cap with white elastomer seal and GL thread.
- Autoclavable (121°C).
Specifications
Polypropylene | |
Soda Lime Glass | |
GL 18 | |
Screw Cap |
160 mm | |
100/Pk. | |
16 mm | |
Culture Tube |
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14
New Offer Save Now -
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%