missing translation for 'onlineSavingsMsg'
Learn More

AccuGENE™ Molecular Biology Water, Lonza™

Product Code. 11558025
Click to view available options
Quantity:
1 L
10 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Ready-to-use for a wide range of molecular biology applications. AccuGENE™ Molecular Biology Water is prepared with 18 megOhm water, filtered using a 0.2 micron filter, and filled into sterile bottles.

  • Reliable - Manufactured according to strict quality control standards to ensure lot-to-lot consistency
  • High Quality - No detectable DNase, RNase, or protease activities
  • Efficient - Ready-made solutions eliminate experiment preparation time
  • Flexible - Customized solutions are available to meet individual needs
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Quantity 10 L
Chemical Name or Material Water

For research or manufacturing use only.

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'